Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158018.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158018.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158018.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158018.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158018.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200158018.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200158018.hg.1 |
TC1200007906.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000300145.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000546709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000549257.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
ENST00000551997.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200007906.hg.1 |
NM_033276.3 |
Refseq |