Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005072.hg.1 |
ENST00000288840.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005072.hg.1 |
ENST00000338426.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005072.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005072.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15005072.hg.1 |
TC15000616.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000558937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000558937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000558937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000558937.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000612349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000612349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000612349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
ENST00000612349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154079.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154079.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154079.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154079.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154079.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154079.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154079.hg.1 |
TC1500007619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500154078.hg.1 |
TC1500007619.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000288840.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000557916.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000558937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000558937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000558937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000558937.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000559931.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000612349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000612349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000612349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
ENST00000612349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NM_005585.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500007619.hg.1 |
NR_027654.1 |
Refseq |