Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Gene 1.0 ST Array |
8067592 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8067589 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8067591 |
- |
- |
Affymetrix Human Gene 1.0 ST Array |
8067590 |
- |
- |
Affymetrix Human Gene 1.1 ST Array |
8067588 |
- |
- |
Affymetrix HT HG-U133+ PM Array Plate |
231760_PM_at |
- |
- |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013994.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013994.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013994.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013994.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013994.hg.1 |
TC20001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013997.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013997.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013997.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013997.hg.1 |
TC20001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013998.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013998.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013998.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013998.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013998.hg.1 |
TC20001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013996.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013996.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013996.hg.1 |
TC20001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013993.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013993.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013993.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013993.hg.1 |
TC20001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013999.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013999.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013999.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20013999.hg.1 |
TC20001023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014000.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014000.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014000.hg.1 |
TC20001023.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009790.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
TC2000009790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168273.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168277.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168277.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168277.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168277.hg.1 |
TC2000009790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168277.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168277.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168277.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168283.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168283.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168283.hg.1 |
TC2000009790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168283.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168283.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
TC2000009790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168271.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
TC2000009790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168272.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
TC2000009790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168279.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
TC2000009790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
ENST00000456634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168281.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
NR_028295.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
TC2000009790.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
ENST00000370341.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
ENST00000414668.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168282.hg.1 |
NR_028295.1 |
Refseq |
Human Genome U133 Plus 2.0 Array |
231760_at |
- |
- |
Human Genome U133 Plus 2.0 Array |
231760_PM_at |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at9 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at7 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at2 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at8 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at11 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at5 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at1 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at6 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at3 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at4 |
- |
- |
Affymetrix Human Genome U133 Plus 2.0 Array |
231760_at10 |
- |
- |
Human Genome U95 Set |
67867_PM_at |
- |
- |
Human Genome U95 Set |
67867_at |
- |
- |
Human Genome U133 Set |
231760_at |
- |
- |
Human Genome U133 Set |
231760_PM_at |
- |
- |