Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009163.hg.1 |
ENST00000262430.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009163.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009163.hg.1 |
TC16000651.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009165.hg.1 |
ENST00000262430.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009165.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009165.hg.1 |
TC16000651.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009164.hg.1 |
ENST00000262430.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009164.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16009164.hg.1 |
TC16000651.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1600191622.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191622.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191622.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191622.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191622.hg.1 |
TC1600011438.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600191623.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191623.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191623.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191623.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191623.hg.1 |
TC1600011438.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
ENST00000569024.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011438.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191625.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191625.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191625.hg.1 |
ENST00000262430.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600191625.hg.1 |
NM_012213.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600191625.hg.1 |
TC1600011438.hg.1 |
Clariom D TC |