Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000261443.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000339438.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000358528.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000369530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000438362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000525878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000534389.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000534699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000610726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001007553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001130523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001242891.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001242892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001242893.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_007158.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000261443.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000339438.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000358528.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000369530.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000438362.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000525878.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000534389.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000534699.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
ENST00000610726.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001007553.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001130523.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001242891.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001242892.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_001242893.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
NM_007158.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100214755.hg.1 |
TC0100015353.hg.1 |
Clariom D TC |