Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381867.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000381873.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
ENST00000614659.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009878.hg.1 |
NM_014723.3 |
Refseq |