Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERKL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030311.2 | NP_001025482.1 | 10 - 11 | 1339 | 60 | |
NM_001030312.2 | NP_001025483.1 | 6 - 7 | 922 | 60 | ||
NM_001030313.2 | NP_001025484.1 | 7 - 8 | 1054 | 60 | ||
NM_001160277.1 | NP_001153749.1 | 9 - 10 | 1207 | 60 | ||
NM_201548.4 | NP_963842.1 | 9 - 10 | 1261 | 60 | ||
NR_027689.1 | 7 - 8 | 1065 | 60 | |||
NR_027690.1 | 8 - 9 | 1197 | 60 | |||
GenBank mRNA | AJ640141.1 | - | 9 - 10 | 1178 | 60 | |
AJ697856.1 | - | 8 - 9 | 1096 | 60 | ||
AK129976.1 | - | 6 - 7 | 504 | 60 | ||
AK293844.1 | - | 9 - 10 | 1208 | 60 | ||
AY357073.1 | - | 9 - 10 | 1160 | 60 | ||
AY690329.1 | - | 10 - 11 | 1238 | 60 | ||
AY690330.1 | - | 6 - 7 | 821 | 60 | ||
AY690331.1 | - | 7 - 8 | 953 | 60 | ||
AY690332.1 | - | 7 - 8 | 964 | 60 | ||
AY690333.1 | - | 8 - 9 | 1096 | 60 | ||
BC137498.1 | - | 9 - 10 | 1187 | 9021498 | 60 | |
BC137499.1 | - | 9 - 10 | 1187 | 9021499 | 60 | |
HQ426666.1 | - | 9 - 10 | 1067 | 60 | ||
HQ426667.1 | - | 8 - 9 | 1003 | 60 | ||
HQ426668.1 | - | 7 - 8 | 871 | 60 | ||
HQ426669.1 | - | 7 - 8 | 860 | 60 | ||
HQ426670.1 | - | 6 - 7 | 728 | 60 | ||
HQ426671.1 | - | 10 - 11 | 1067 | 60 | ||
HQ426672.1 | - | 9 - 10 | 989 | 60 | ||
HQ426673.1 | - | 8 - 9 | 925 | 60 | ||
HQ426674.1 | - | 7 - 8 | 793 | 60 | ||
HQ426675.1 | - | 7 - 8 | 782 | 60 | ||
HQ426676.1 | - | 6 - 7 | 650 | 60 | ||
HQ426677.1 | - | 7 - 8 | 597 | 60 | ||
HQ426678.1 | - | 10 - 11 | 1314 | 60 | ||
HQ426679.1 | - | 7 - 8 | 1619 | 60 | ||
JF290420.1 | - | 9 - 10 | 1147 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |