Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025806.hg.1 |
TC01001667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025806.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025806.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025807.hg.1 |
TC01001667.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025807.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01025807.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182118.hg.1 |
TC0100011200.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100182117.hg.1 |
TC0100011200.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000367287.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000478378.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000482943.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
ENST00000484647.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100011200.hg.1 |
NM_006335.2 |
Refseq |