Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010983.hg.1 |
ENST00000216085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010983.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010983.hg.1 |
TC22000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010984.hg.1 |
ENST00000216085.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010984.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR22010984.hg.1 |
TC22000632.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007012.hg.1 |
ENST00000433125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200007012.hg.1 |
ENST00000433125.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000406335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000406335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000406335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000406335.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160028.hg.1 |
TC2200008393.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160028.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160028.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160028.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160028.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160028.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160028.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
TC2200008393.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2200160029.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200007012.hg.1 |
ENST00000433125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200007012.hg.1 |
ENST00000433125.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000216085.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000406335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000406335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000406335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000406335.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000413137.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000414672.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000488106.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000493894.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
ENST00000496342.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2200008393.hg.1 |
NM_012265.2 |
Refseq |