Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015104.2 | NP_055919.2 | 37 - 38 | 5269 | 63 | |
XM_005273849.3 | XP_005273906.1 | 37 - 38 | 5242 | 63 | ||
XM_005273850.3 | XP_005273907.1 | 37 - 38 | 5236 | 63 | ||
XM_011544863.2 | XP_011543165.1 | 37 - 38 | 5266 | 63 | ||
XM_011544864.2 | XP_011543166.1 | 37 - 38 | 5251 | 63 | ||
XM_011544865.2 | XP_011543167.1 | 37 - 38 | 5259 | 63 | ||
XM_011544866.2 | XP_011543168.1 | 37 - 38 | 5266 | 63 | ||
XM_011544868.2 | XP_011543170.1 | 22 - 23 | 2880 | 63 | ||
GenBank mRNA | AB007864.1 | - | 37 - 38 | 5210 | 63 | |
BC008593.1 | - | 8 - 9 | 897 | 63 | ||
BC053596.1 | - | 12 - 13 | 1597 | 63 | ||
BC110650.1 | - | 37 - 38 | 5269 | 6147561 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |