Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIK3CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006219.2 | NP_006210.1 | 4 - 5 | 814 | 87 | |
XM_005247530.2 | XP_005247587.1 | 6 - 7 | 1163 | 87 | ||
XM_006713659.3 | XP_006713722.1 | 5 - 6 | 1034 | 87 | ||
XM_011512895.2 | XP_011511197.1 | 7 - 8 | 1159 | 87 | ||
XM_011512896.1 | XP_011511198.1 | 3 - 4 | 479 | 87 | ||
XM_017006619.1 | XP_016862108.1 | 5 - 6 | 837 | 87 | ||
XM_017006620.1 | XP_016862109.1 | 3 - 4 | 479 | 87 | ||
XM_017006621.1 | XP_016862110.1 | 3 - 4 | 479 | 87 | ||
GenBank mRNA | BC114432.1 | - | 4 - 5 | 802 | 40008544 | 87 |
S67334.1 | - | 4 - 5 | 798 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons |