Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024642.hg.1 |
TC05001816.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024642.hg.1 |
ENST00000282223.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024642.hg.1 |
ENST00000394945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024642.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024642.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024643.hg.1 |
TC05001816.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024643.hg.1 |
ENST00000282223.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024643.hg.1 |
ENST00000394945.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024643.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05024643.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000509293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000509293.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000503916.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000505690.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000509293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000509293.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510405.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000510689.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
ENST00000515091.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500012138.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183984.hg.1 |
TC0500012138.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
ENST00000282223.11 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
ENST00000394945.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
ENST00000508642.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
ENST00000509978.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
NM_004598.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500183985.hg.1 |
TC0500012138.hg.1 |
Clariom D TC |