Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012711.hg.1 |
TC05000868.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012711.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012711.hg.1 |
ENST00000521390.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05012711.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500164567.hg.1 |
TC0500009215.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000311946.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000435489.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000519946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
ENST00000521390.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001099287.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500009215.hg.1 |
NM_001172292.1 |
Refseq |