Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002098.hg.1 |
ENST00000321341.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002098.hg.1 |
ENST00000382927.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002098.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002098.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR18002098.hg.1 |
TC18000180.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000591925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000591925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
ENST00000591925.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
TC1800007326.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1800150096.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000321341.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000382927.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000585680.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000589330.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000591925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000591925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
ENST00000591925.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_001168335.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1800007326.hg.1 |
NM_002396.4 |
Refseq |