Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005924.hg.1 |
ENST00000380752.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005924.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13005924.hg.1 |
TC13000523.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000450494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000450494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000450494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000450494.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000473577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000473577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000473577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
ENST00000473577.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000450494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000450494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000450494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000450494.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000473577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000473577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000473577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
ENST00000473577.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300008487.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157219.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157219.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157219.hg.1 |
ENST00000380752.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300157219.hg.1 |
NM_003045.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300157219.hg.1 |
TC1300008487.hg.1 |
Clariom D TC |