Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM69 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001301144.1 | NP_001288073.1 | 4 - 5 | 713 | 136 | |
NM_001301145.1 | NP_001288074.1 | 4 - 5 | 662 | 85 | ||
NM_080745.4 | NP_542783.2 | 4 - 5 | 705 | 136 | ||
NM_182985.4 | NP_892030.3 | 5 - 6 | 1182 | 136 | ||
GenBank mRNA | AF302088.1 | - | 4 - 5 | 561 | 136 | |
AF302089.1 | - | 4 - 5 | 510 | 85 | ||
AK226115.1 | - | 3 - 4 | 4969 | 136 | ||
AK292252.1 | - | 5 - 6 | 1026 | 136 | ||
AL360161.1 | - | 4 - 5 | 548 | 136 | ||
AY305385.1 | - | 5 - 6 | 880 | 136 | ||
BC024199.1 | - | 5 - 6 | 1000 | 136 | ||
BC033314.1 | - | 4 - 5 | 501 | 5166812 | 136 | |
BC047945.1 | - | 5 - 6 | 1019 | 5744224 | 136 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |