Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195059.1 | NP_001181988.1 | 1 - 2 | 147 | 59 | |
GenBank mRNA | BC008744.2 | - | 1 - 2 | 108 | 3346334 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |