Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015918.hg.1 |
ENST00000228434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015918.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015918.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015918.hg.1 |
TC12001207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015917.hg.1 |
ENST00000228434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015917.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015917.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015917.hg.1 |
TC12001207.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015919.hg.1 |
ENST00000228434.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015919.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015919.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015919.hg.1 |
TC12001207.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174412.hg.1 |
TC1200009876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174414.hg.1 |
TC1200009876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174410.hg.1 |
TC1200009876.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1200009876.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
ENST00000228434.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
ENST00000416624.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
ENST00000536709.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
ENST00000543147.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
NM_001781.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200174413.hg.1 |
TC1200009876.hg.1 |
Clariom D TC |