Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTPRK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001135648.2 | NP_001129120.1 | 18 - 19 | 3163 | 111 | |
NM_001291981.1 | NP_001278910.1 | 20 - 21 | 3211 | 111 | ||
XM_006715537.2 | XP_006715600.1 | 18 - 19 | 3211 | 111 | ||
XM_011536014.2 | XP_011534316.1 | 20 - 21 | 3259 | 111 | ||
XM_011536015.2 | XP_011534317.1 | 19 - 20 | 3250 | 111 | ||
XM_011536017.2 | XP_011534319.1 | 21 - 22 | 3502 | 111 | ||
XM_011536018.2 | XP_011534320.1 | 21 - 22 | 3175 | 111 | ||
XM_017011145.1 | XP_016866634.1 | 19 - 20 | 3178 | 111 | ||
XM_017011146.1 | XP_016866635.1 | 19 - 20 | 3127 | 111 | ||
XM_017011147.1 | XP_016866636.1 | 19 - 20 | 3105 | 111 | ||
XM_017011149.1 | XP_016866638.1 | 8 - 9 | 763 | 111 | ||
XM_017011150.1 | XP_016866639.1 | 7 - 8 | 750 | 111 | ||
GenBank mRNA | BC140775.1 | - | 18 - 19 | 2965 | 9021643 | 111 |
BC144512.1 | - | 18 - 19 | 2965 | 9053039 | 111 | |
BC144513.1 | - | 20 - 21 | 3013 | 9053040 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Protein Phosphatase Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |