Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002164.hg.1 |
TC05000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002164.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002164.hg.1 |
ENST00000440677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002164.hg.1 |
ENST00000515157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002164.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002165.hg.1 |
TC05000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002165.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002165.hg.1 |
ENST00000440677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002165.hg.1 |
ENST00000515157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002165.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000486779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000486779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000486779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000486779.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000489457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000489457.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000494960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000494960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000494960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000494960.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000508185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000508185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000508185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000508185.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000513922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000513922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
ENST00000513922.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002163.hg.1 |
TC05000187.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002163.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002163.hg.1 |
ENST00000440677.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002163.hg.1 |
ENST00000515157.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR05002163.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148884.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148884.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148884.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148884.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148884.hg.1 |
TC0500007238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0500148881.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148881.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148881.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0500148881.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0500148881.hg.1 |
TC0500007238.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000313164.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000464864.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000486779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000486779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000486779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000486779.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000489457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000489457.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000494960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000494960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000494960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000494960.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000508185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000508185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000508185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000508185.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000513922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000513922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
ENST00000513922.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500007238.hg.1 |
NM_000587.3 |
Refseq |