Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
ENST00000263574.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
ENST00000278756.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
ENST00000338167.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
ENST00000345598.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
ENST00000528499.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
ENST00000539648.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
ENST00000543137.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
NM_001142276.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
NM_001142277.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
NM_001142278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
NM_001243299.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
NM_001642.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
NR_024515.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
NR_024516.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11015006.hg.1 |
TC11001194.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000263574.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000278756.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000338167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000345598.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000528499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000533616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001142276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001142277.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001142278.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001243299.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001642.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NR_024515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NR_024516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000263574.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000278756.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000338167.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000345598.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000528499.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
ENST00000533616.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001142276.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001142277.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001142278.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001243299.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NM_001642.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NR_024515.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
NR_024516.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100171575.hg.1 |
TC1100009521.hg.1 |
Clariom D TC |