Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006013.hg.1 |
TC0X000468.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006013.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006013.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006014.hg.1 |
TC0X000468.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006014.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X006014.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153748.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153748.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153748.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153748.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153748.hg.1 |
TC0X00007918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0X00153747.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153747.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153747.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153747.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153747.hg.1 |
TC0X00007918.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
ENST00000485971.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0X00007918.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153749.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153749.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153749.hg.1 |
ENST00000373031.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00153749.hg.1 |
NM_022144.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00153749.hg.1 |
TC0X00007918.hg.1 |
Clariom D TC |