Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001037.hg.1 |
ENST00000378845.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001037.hg.1 |
ENST00000378847.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001037.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001037.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001037.hg.1 |
TC10000093.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000487696.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006802.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
ENST00000378845.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
ENST00000615792.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
ENST00000619168.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
NM_020397.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
NM_153498.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000146499.hg.1 |
TC1000006802.hg.1 |
Clariom D TC |