Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015525.hg.1 |
TC09001211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015525.hg.1 |
ENST00000297785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015525.hg.1 |
ENST00000428593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015525.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015524.hg.1 |
TC09001211.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015524.hg.1 |
ENST00000297785.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015524.hg.1 |
ENST00000376939.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015524.hg.1 |
ENST00000428593.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR09015524.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186218.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186218.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186218.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186218.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186218.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186218.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186218.hg.1 |
TC0900012245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0900186219.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186219.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186219.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0900186219.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0900186219.hg.1 |
TC0900012245.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000297785.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000376939.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000446946.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000482210.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
ENST00000493113.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0900012245.hg.1 |
NM_000689.4 |
Refseq |