Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057922.hg.1 |
TC01003778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057922.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057922.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000460870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000460870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000460870.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000463510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000463510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000463510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000463510.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000480053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000480053.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000486969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000486969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
ENST00000486969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057921.hg.1 |
TC01003778.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057921.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01057921.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230009.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230009.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230009.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230009.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230009.hg.1 |
TC0100017145.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100230010.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230010.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230010.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100230010.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100230010.hg.1 |
TC0100017145.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000367033.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000460870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000460870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000460870.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000463510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000463510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000463510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000463510.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000480053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000480053.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000483048.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000486969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000486969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
ENST00000486969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100017145.hg.1 |
NM_025179.3 |
Refseq |