Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0300011236.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
ENST00000273286.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
NM_020678.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184131.hg.1 |
TC0300011236.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0300184135.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184135.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184135.hg.1 |
ENST00000493075.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0300184135.hg.1 |
NM_001304389.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0300184135.hg.1 |
TC0300011236.hg.1 |
Clariom D TC |