Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017225.hg.1 |
TC01001094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017225.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017225.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017227.hg.1 |
TC01001094.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017227.hg.1 |
ENST00000369306.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017227.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017227.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR01017227.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000428634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000428634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000428634.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242795.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242795.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242795.hg.1 |
TC0100018476.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100242798.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242798.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242798.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242798.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242798.hg.1 |
TC0100018476.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100242792.hg.1 |
TC0100018476.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000369306.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000428634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000428634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000428634.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
ENST00000537888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_001184795.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NM_003846.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073491.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073492.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100018476.hg.1 |
NR_073493.2 |
Refseq |