Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054344.hg.1 |
TC01003497.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054344.hg.1 |
ENST00000236137.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054344.hg.1 |
ENST00000367802.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054344.hg.1 |
ENST00000367804.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR01054344.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224344.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224344.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224344.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224344.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224344.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0100224344.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0100224344.hg.1 |
TC0100016351.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000236137.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
ENST00000367804.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0100016351.hg.1 |
NM_006996.2 |
Refseq |