Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001622.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001622.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10001622.hg.1 |
TC10000140.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000006924.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147421.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147421.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147421.hg.1 |
ENST00000377275.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000147421.hg.1 |
NM_178815.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000147421.hg.1 |
TC1000006924.hg.1 |
Clariom D TC |