Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010206.hg.1 |
ENST00000278980.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010206.hg.1 |
ENST00000446419.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010206.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010206.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20010206.hg.1 |
TC20000766.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
TC2000008860.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000161760.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000278980.10 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000446419.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000474815.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
ENST00000610160.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_001099339.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000008860.hg.1 |
NM_053041.2 |
Refseq |