Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014347.hg.1 |
ENST00000369886.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014347.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014347.hg.1 |
TC20001044.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014345.hg.1 |
ENST00000369886.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014345.hg.1 |
ENST00000498830.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014345.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR20014345.hg.1 |
TC20001044.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
TC2000009851.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168827.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168829.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168829.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168829.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168829.hg.1 |
TC2000009851.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168829.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168829.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168829.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168830.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168830.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168830.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2000168830.hg.1 |
TC2000009851.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2000168830.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168830.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2000168830.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000369886.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000450107.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000478694.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
ENST00000498830.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC2000009851.hg.1 |
NM_080621.4 |
Refseq |