Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VN1R10P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_045612.1 | 1 | 983 | 161 | ||
GenBank mRNA | AF336874.1 | - | 1 | 944 | 161 | |
AF352327.1 | - | 1 | 962 | 161 | ||
BC107050.2 | - | 1 | 945 | 40004452 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens 1700 Both primers and probe map within a single exon |