Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018253.hg.1 |
ENST00000393246.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018253.hg.1 |
ENST00000539556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018253.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018253.hg.1 |
TC16001316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018252.hg.1 |
ENST00000393246.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018252.hg.1 |
ENST00000539556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018252.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018252.hg.1 |
TC16001316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018251.hg.1 |
ENST00000393246.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018251.hg.1 |
ENST00000539556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018251.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018251.hg.1 |
TC16001316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018254.hg.1 |
ENST00000393246.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018254.hg.1 |
ENST00000539556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018254.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018254.hg.1 |
TC16001316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018250.hg.1 |
ENST00000393246.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018250.hg.1 |
ENST00000539556.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018250.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16018250.hg.1 |
TC16001316.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186132.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186132.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186132.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186132.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186132.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186132.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186132.hg.1 |
TC1600011080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186130.hg.1 |
TC1600011080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600186136.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186136.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186136.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186136.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186136.hg.1 |
TC1600011080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600186135.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186135.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186135.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186135.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186135.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186135.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186135.hg.1 |
TC1600011080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600011080.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
ENST00000618669.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186131.hg.1 |
TC1600011080.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1600186133.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186133.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186133.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186133.hg.1 |
ENST00000539556.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186133.hg.1 |
ENST00000629253.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600186133.hg.1 |
NM_198491.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600186133.hg.1 |
TC1600011080.hg.1 |
Clariom D TC |