Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEF2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005920.3 | NP_005911.1 | 8 - 9 | 1371 | 81 | |
XM_005245169.4 | XP_005245226.1 | 8 - 9 | 1353 | 81 | ||
XM_005245170.3 | XP_005245227.1 | 8 - 9 | 1378 | 81 | ||
XM_006711330.3 | XP_006711393.1 | 8 - 9 | 1082 | 81 | ||
XM_006711332.3 | XP_006711395.1 | 8 - 9 | 1366 | 81 | ||
XM_011509569.2 | XP_011507871.1 | 8 - 9 | 1403 | 81 | ||
GenBank mRNA | AB210005.1 | - | 8 - 9 | 1285 | 81 | |
AK308641.1 | - | 8 - 9 | 1271 | 81 | ||
BC054520.1 | - | 8 - 9 | 1266 | 6142751 | 81 | |
L16794.1 | - | 8 - 9 | 1071 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons |