Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007780.hg.1 |
TC06000640.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007780.hg.1 |
ENST00000335783.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007780.hg.1 |
ENST00000371200.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06007780.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157419.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157419.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157419.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600157419.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600157419.hg.1 |
TC0600008209.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
ENST00000335783.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600008209.hg.1 |
NM_018132.3 |
Refseq |