Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THSD7A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015204.2 | NP_056019.1 | 20 - 21 | 4164 | 97 | |
XM_006715659.1 | XP_006715722.1 | 20 - 21 | 4163 | 97 | ||
XM_006715660.1 | XP_006715723.1 | 20 - 21 | 4163 | 97 | ||
XM_006715662.1 | XP_006715725.1 | 20 - 21 | 4163 | 97 | ||
XM_011515193.2 | XP_011513495.1 | 20 - 21 | 6488 | 97 | ||
XM_011515194.1 | XP_011513496.1 | 20 - 21 | 3929 | 97 | ||
GenBank mRNA | AB023177.2 | - | 19 - 20 | 3448 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons |