Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014450.hg.1 |
ENST00000397801.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014450.hg.1 |
ENST00000538940.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014450.hg.1 |
NM_022370.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11014450.hg.1 |
TC11001146.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000397801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000525448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000526551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000527196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000527245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000528068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000528820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000530647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000531075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000531119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000531545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000531888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000534598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000538940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
NM_022370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000397801.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000525448.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000526551.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000527196.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000527245.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000528068.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000528820.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000530647.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000531075.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000531119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000531545.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000531888.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000534598.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
ENST00000538940.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
NM_022370.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100170471.hg.1 |
TC1100009403.hg.1 |
Clariom D TC |