Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007636.hg.1 |
ENST00000512867.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0800007636.hg.1 |
ENST00000512867.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558100.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000558891.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000560774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000560774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
ENST00000560774.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003886.hg.1 |
ENST00000261845.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003886.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR15003886.hg.1 |
TC15000417.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007593.hg.1 |
ENST00000420752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007593.hg.1 |
ENST00000420752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007593.hg.1 |
ENST00000420752.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000007593.hg.1 |
ENST00000420752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558100.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558841.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000558891.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000560774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000560774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
ENST00000560774.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1500010743.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0800007636.hg.1 |
ENST00000512867.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0800007636.hg.1 |
ENST00000512867.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182454.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182454.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182454.hg.1 |
ENST00000261845.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1500182454.hg.1 |
NM_002748.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1500182454.hg.1 |
TC1500010743.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1000007593.hg.1 |
ENST00000420752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007593.hg.1 |
ENST00000420752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007593.hg.1 |
ENST00000420752.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000007593.hg.1 |
ENST00000420752.1 |
Ensembl |