Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000502722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000502722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000502722.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000514587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000514587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000514587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000514587.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
ENST00000515349.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000380345.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000502722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000502722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000502722.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000503686.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000510949.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000512759.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000514587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000514587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000514587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000514587.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515007.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
ENST00000515349.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0500011225.hg.1 |
NM_005779.2 |
Refseq |