Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011974.hg.1 |
ENST00000330063.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011974.hg.1 |
ENST00000399609.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011974.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011974.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR16011974.hg.1 |
TC16000825.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1600009217.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
ENST00000399609.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
ENST00000537996.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
ENST00000572111.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
NM_001098814.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1600170458.hg.1 |
TC1600009217.hg.1 |
Clariom D TC |