Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030283.hg.1 |
TC07001965.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030283.hg.1 |
ENST00000355265.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07030283.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000460479.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000465697.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000467543.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000470850.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000476829.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000479768.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
ENST00000494148.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0700012870.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700192871.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192871.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192871.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700192871.hg.1 |
ENST00000355265.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192871.hg.1 |
ENST00000478969.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700192871.hg.1 |
NM_000420.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700192871.hg.1 |
TC0700012870.hg.1 |
Clariom D TC |