Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005957.hg.1 |
ENST00000262631.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005957.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR19005957.hg.1 |
TC19000455.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000601692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000601692.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000415950.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000596348.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000601692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000601692.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1900007830.hg.1 |
NM_199037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
TC1900007830.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157724.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
NM_001037.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
TC1900007830.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
ENST00000262631.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
ENST00000595652.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
ENST00000602150.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1900157722.hg.1 |
NM_001037.4 |
Refseq |