Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABBR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021904.3 | NP_068704.2 | 4 - 5 | 602 | 65 | |
XM_006715047.3 | XP_006715110.1 | 4 - 5 | 403 | 65 | ||
GenBank mRNA | AJ012187.1 | - | 3 - 4 | 316 | 65 | |
AL539611.3 | - | 4 - 5 | 602 | 65 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 G-protein coupled receptor Homo sapiens Probe spans exons |