Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
ENST00000300527.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
ENST00000310645.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
ENST00000357838.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
ENST00000397763.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
ENST00000409416.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
NM_001849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
NM_058174.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
NM_058175.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR21003101.hg.1 |
TC21000253.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000300527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000310645.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000397763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000409416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000413758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
NM_001849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
NM_058174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
NM_058175.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
TC2100007451.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000300527.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000310645.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000397763.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000409416.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
ENST00000413758.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
NM_001849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
NM_058174.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR2100150540.hg.1 |
NM_058175.2 |
Refseq |