Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000335.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000335.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000335.hg.1 |
TC11000023.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000334.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000334.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000334.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11000334.hg.1 |
TC11000023.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1100144672.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144672.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144672.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144672.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144672.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144672.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144672.hg.1 |
TC1100006481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100144673.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144673.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144673.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144673.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144673.hg.1 |
TC1100006481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1100144671.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144671.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144671.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144671.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144671.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100144671.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100144671.hg.1 |
TC1100006481.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000176183.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
ENST00000528733.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100006481.hg.1 |
NM_000797.3 |
Refseq |