Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009624.hg.1 |
ENST00000245323.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009624.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR13009624.hg.1 |
TC13000860.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164884.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164884.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164884.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1300164884.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1300164884.hg.1 |
TC1300009712.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
ENST00000245323.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1300009712.hg.1 |
NM_004093.3 |
Refseq |