Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USPL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_005800.4 | NP_005791.3 | 2 - 3 | 537 | 81 | |
XM_005266213.3 | XP_005266270.1 | 3 - 4 | 895 | 81 | ||
XM_017020314.1 | XP_016875803.1 | 2 - 3 | 833 | 81 | ||
GenBank mRNA | AK289826.1 | - | 2 - 3 | 293 | 81 | |
BC036027.1 | - | 2 - 3 | 309 | 5271146 | 81 | |
BC038103.2 | - | 2 - 3 | 447 | 4821753 | 81 | |
X59131.1 | - | 2 - 3 | 282 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |