Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_006289.3 | NP_006280.3 | 39 - 40 | 5544 | 111 | |
GenBank mRNA | AB028950.2 | - | 39 - 40 | 5519 | 111 | |
AF078828.1 | - | 39 - 40 | 5316 | 111 | ||
AF177198.1 | - | 39 - 40 | 5280 | 111 | ||
BC042923.1 | - | 39 - 40 | 5330 | 5576203 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons |