Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OTOGL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173591.3 | NP_775862.3 | 37 - 38 | 4401 | 149 | |
XM_005268802.3 | XP_005268859.1 | 41 - 42 | 5202 | 149 | ||
XM_011538191.1 | XP_011536493.1 | 38 - 39 | 4478 | 149 | ||
XM_011538192.2 | XP_011536494.1 | 35 - 36 | 4419 | 149 | ||
XM_011538193.1 | XP_011536495.1 | 35 - 36 | 4338 | 149 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons |