Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001207047.2 | NP_001193976.1 | 3 - 4 | 314 | 63 | |
NM_001323332.1 | NP_001310261.1 | 3 - 4 | 697 | 63 | ||
NM_139321.2 | NP_647537.1 | 3 - 4 | 697 | 63 | ||
NM_139322.3 | NP_647538.1 | 3 - 4 | 697 | 63 | ||
GenBank mRNA | AF106861.1 | - | 3 - 4 | 607 | 63 | |
AK293010.1 | - | 3 - 4 | 697 | 63 | ||
AK302730.1 | - | 3 - 4 | 293 | 63 | ||
BC101705.1 | - | 3 - 4 | 647 | 8069211 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons |